missing translation for 'onlineSavingsMsg'
Learn More

Acetamide 98.0+%, TCI America™

Catalog No. p-7116653
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
A000725G 25 g
A0007500G 500 g
2 options

Catalog No. A000725G

Supplier: TCI America A000725G

Only null left

Chemical Identifiers

CAS 60-35-5
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.068
MDL Number MFCD00008023
InChI Key DLFVBJFMPXGRIB-UHFFFAOYSA-N
Synonym ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
PubChem CID 178
ChEBI CHEBI:49028
IUPAC Name acetamide
SMILES CC(=O)N

Specifications

Melting Point 81°C
Color White
Boiling Point 222°C
Quantity 25 g
Formula Weight 59.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acetamide
EINECSNumber (2)-0724
RTECSNumber AB4025000
TSCA Yes