missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, 99%

CAS: 60-35-5 | C2H5NO | 59.068 g/mol

Supplier:  Thermo Scientific Chemicals A125890B

 View more versions of this product

Catalog No. AAA125890B


Only null left
Explore available promotions

Description

Description

Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
Specifications

Specifications

Acetamide
76°C to 81°C
220°C to 222°C
MFCD00008023
1071207
14,43
Soluble in water.
CC(=O)N
59.068
CHEBI:49028
99%
60-35-5
1.159
C2H5NO
1000 g
Hygroscopic
ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
DLFVBJFMPXGRIB-UHFFFAOYSA-N
acetamide
178
59.07

Chemical Identifiers

60-35-5
59.068
DLFVBJFMPXGRIB-UHFFFAOYSA-N
178
acetamide
C2H5NO
MFCD00008023
ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
CHEBI:49028
CC(=O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

P201-P202-P281-P308+P313-P501c

H351

EINECSNumber : 200-473-5

RTECSNumber : AB4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only