missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, 99%, for analysis

Catalog No. AC451870010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451870010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC451870010 Supplier Thermo Scientific Chemicals Supplier No. 451870010
Only null left

CAS: 60-35-5 | C2H5NO | 59.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-35-5
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.06
InChI Key DLFVBJFMPXGRIB-UHFFFAOYSA-N
Synonym ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
PubChem CID 178
ChEBI CHEBI:49028
IUPAC Name acetamide
SMILES CC(=O)N

Specifications

Melting Point 79.0°C to 81.0°C
Boiling Point 221.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3CONH2
Quantity 1 kg
Merck Index 15,44
Solubility Information Solubility in water: 2000g/L (20°C). Other solubilities: 1 g/2 mL alcohol - 1 g/6 mL pyridine, soluble in chloroform, glycerol and hot benzene, slightly soluble in ether
Formula Weight 59.06
Percent Purity 99%
Grade Analysis
Insoluble Matter 0.025% max. (in water)
Chemical Name or Material Acetamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 200-473-5

RUO – Research Use Only