missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde, Reagent, ACS, 99.5%, Spectrum™ Chemical

Catalog No. p-7156966
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600184 100 mL Amber Glass Bottle
18600185 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600184 Supplier Spectrum Chemical Mfg Cor Supplier No. A2147100ML
Only null left

A2147, 75-07-0, C2H4O

Spectrum™ Chemical Acetaldehyde, Reagent, ACS is commercially used as a precursor to acetic acid. In the laboratory it is a reagent used for the detection of primary and secondary amines. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 75-07-0
Molecular Formula C2H4O
Molecular Weight (g/mol) 44.05
InChI Key IKHGUXGNUITLKF-UHFFFAOYSA-N
IUPAC Name acetaldehyde
SMILES CC=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
UN Number UN1089
Residue after Evaporation 0.005%
Percent Purity 99.5%
Grade ACS