missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde Ethyl cis-3-Hexenyl Acetal 97.0+%, TCI America™

Catalog No. A092825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A092825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A092825ML Supplier TCI America Supplier No. A092825ML
Only null left

Chemical Identifiers

CAS 28069-74-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00036618
InChI Key PAEBAEDUARAOSG-VOTSOKGWSA-N
PubChem CID 5463962
IUPAC Name (E)-1-(1-ethoxyethoxy)hex-3-ene
SMILES CCC=CCCOC(C)OCC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Acetaldehyde Ethyl cis-3-Hexenyl Acetal
EINECSNumber (2)-2458
RTECSNumber MP6795000
TSCA Yes