missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetaldehyde diethyl acetal, 99%
Description
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.
Solubility
Soluble in water (46 g/L at 20°C).
Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with oxidizing agents.
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.
Solubility
Soluble in water (46 g/L at 20°C).
Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -100°C |
| Density | 0.831 |
| Boiling Point | 101°C to 102°C |
| Flash Point | −21°C (−6°F) |
| Odor | Pleasant |
| Refractive Index | 1.381 |
| Quantity | 100 g |
| UN Number | UN1088 |
| Beilstein | 1098310 |
| Merck Index | 14,38 |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319 |
| Hazard Statement | GHS H Statement H225-H315-H319 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETAL |
| EINECSNumber | 203-310-6 |
| RTECSNumber | AB2800000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only