missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde diethyl acetal, 99%

Catalog No. AAA1370136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1370136 500 g
AAA1370122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1370136 Supplier Thermo Scientific Chemicals Supplier No. A1370136

CAS: 105-57-7 | C6H14O2 | 118.18 g/mol

Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.

Solubility
Soluble in water (46 g/L at 20°C).

Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 105-57-7
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009243
InChI Key DHKHKXVYLBGOIT-UHFFFAOYSA-N
Synonym acetal, acetaldehyde diethyl acetal, diethyl acetal, ethane, 1,1-diethoxy, diaethylacetal, acetale, ethylidene diethyl ether, acetal diethylique, diethylacetal, 1,1-dietossietano
PubChem CID 7765
IUPAC Name 1,1-diethoxyethane
SMILES CCOC(C)OCC

Specifications

Melting Point -100°C
Density 0.831
Boiling Point 101°C to 102°C
Flash Point −21°C (−6°F)
Odor Pleasant
Refractive Index 1.381
Quantity 500 g
UN Number UN1088
Beilstein 1098310
Merck Index 14,38
Solubility Information Soluble in water (46g/L at 20°C).
Formula Weight 118.18
Percent Purity 99%
Chemical Name or Material Acetaldehyde diethyl acetal
Show More Show Less
Hazard Category H225-H315-H319
Hazard Statement GHS H Statement
H225-H315-H319
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETAL
EINECSNumber 203-310-6
RTECSNumber AB2800000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only