missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde (ca. 2% in N,N-Dimethylformamide) [for Detection of Primary and Secondary Amines], TCI America™

Supplier:  TCI America A164010ML

Catalog No. A164010ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetaldehyde (ca. 2% in N,N-Dimethylformamide) [for Detection of Primary and Secondary Amines]
Yellow
MFCD00006991
1992
IKHGUXGNUITLKF-UHFFFAOYSA-N
acetaldehyde
177
44.05
75-07-0
C2H4O
10 mL
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
CC=O
44.053
CHEBI:15343
Liquid

Chemical Identifiers

75-07-0
44.053
IKHGUXGNUITLKF-UHFFFAOYSA-N
177
acetaldehyde
C2H4O
MFCD00006991
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
CHEBI:15343
CC=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0485

RTECSNumber : AB1925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions