missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde Ammonia Trimer 95.0+%, TCI America™

Catalog No. A020125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A020125G 25 g
A0201500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A020125G Supplier TCI America Supplier No. A020125G

Chemical Identifiers

CAS 58052-80-5
Molecular Formula C6H18N3
Molecular Weight (g/mol) 132.23
MDL Number MFCD00149559
InChI Key MZSSRMMSFLVKPK-UHFFFAOYSA-Q
Synonym 2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt
PubChem CID 2723814
IUPAC Name 2,4,6-trimethyl-1,3,5-triazinane-1,3,5-triium
SMILES CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 25 g
UN Number 1841
Formula Weight 129.21
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetaldehyde Ammonia Trimer
EINECSNumber (2)-0344
RTECSNumber AB1950000
TSCA No