missing translation for 'onlineSavingsMsg'
Learn More

Acetal 98.0+%, TCI America™

Catalog No. D045525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D045525ML 25 mL
D0455500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D045525ML Supplier TCI America Supplier No. D045525ML

Chemical Identifiers

CAS 105-57-7
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009243
InChI Key DHKHKXVYLBGOIT-UHFFFAOYSA-N
Synonym acetal, acetaldehyde diethyl acetal, diethyl acetal, ethane, 1,1-diethoxy, diaethylacetal, acetale, ethylidene diethyl ether, acetal diethylique, diethylacetal, 1,1-dietossietano
PubChem CID 7765
IUPAC Name 1,1-diethoxyethane
SMILES CCOC(C)OCC

Specifications

Melting Point -100°C
Color Colorless
Boiling Point 103°C
Quantity 25 mL
UN Number 1088
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Acetal
EINECSNumber (2)-0496
RTECSNumber AB2800000
TSCA Yes