Learn More
Cytiva POM Column Clamp
Catalog No. 361041186
Change view
Click to view available options
For Use With:
ÄKTA Avant, ÄKTA Pure Using The Rail System
Catalog No. For Use With
361041186 ÄKTA Avant, ÄKTA Pure Using The Rail System
1 options
Catalog No. 361041186 Supplier Cytiva Supplier No. 28956319
Only null left
For use with ÄKTA™ Avant and Pure chromatography systems. Cytiva POM Column Clamp is an acetal plastic (POM) holder that easily inserts into an ÄKTA rail system for holding columns with a 10-21mm O.D.
  • Pack of five (5) column clamps that hold columns with 10–21mm outer diameter
  • Easily attaches to ÄKTA™ Avant or ÄKTA™ Pure instruments using the rail system:
    • Press inner end tabs of the clamp together
    • Insert pressed end into the rail of the instrument
    • Turn the clamp 90° and release
Specifications
Type Clamp
Diameter (Metric) Outer For 10 to 21 mm OD columns
Packaging Quantity 5 Pack
For Use With ÄKTA Avant, ÄKTA Pure Using The Rail System
Material POM