missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame Potassium, NF, 99-101%, Spectrum™ Chemical

Catalog No. 18600181
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600181 1 kg Amber Glass Bottle
18600182 25 g Amber Glass Bottle
18600180 100 g Amber Glass Bottle
18600183 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600181 Supplier Spectrum Chemical Mfg Cor Supplier No. A39741KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A3974, 55589-62-3, C4H4KNO4S

Spectrum™ Chemical Acesulfame Potassium, NF is a white crystallized powder and is a sugar substitute that has no calories. It is 200 times sweeter than sugar and stable under heat and used commercially as a sweetener in baking and in products that need to have a long shelf life. It is also used in combination with other sweeteners in carbonated drinks and is typically used as the main sweetening agent in pharmaceutical products. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 55589-62-3
Molecular Formula C4H4KNO4S
Molecular Weight (g/mol) 201.24
InChI Key JLEKLYQXZHJOTQ-UHFFFAOYSA-M
IUPAC Name potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
SMILES [K+].CC1=CC(=O)N=S([O-])(=O)O1

Specifications

CAS Max % 100%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 99 to 101%
Grade NF