missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acesulfame Potassium, 99%, Spectrum™ Chemical
Description
Spectrum™ Chemical Acesulfame Potassium is a sugar substitute that has no calories and is a white crystalline powder. Thanks to the fact that it is 200 times sweeter than sugar and stable under heat, it is used commercially as a sweetener in baking and products that need to have a long shelf life. It is also used in combination with other sweeteners in carbonated drinks and is typically used as the main sweetening agent in pharmaceutical products.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Pail |
| Quantity | 25 kg |
| Percent Purity | 99% |
| Grade | Reagent |