missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame Potassium, 99%, Spectrum™ Chemical

Catalog No. 18600169
Change view
Click to view available options
Quantity:
25 g
1 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600169 25 g Amber Glass Bottle
18600171 1 kg Amber Glass Bottle
18600173 12 kg Poly Pail
18600172 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600169 Supplier Spectrum Chemical Mfg Cor Supplier No. A281525GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A2815, 55589-62-3, C4H4KNO4S

Spectrum™ Chemical Acesulfame Potassium is a sugar substitute that has no calories and is a white crystalline powder. Thanks to the fact that it is 200 times sweeter than sugar and stable under heat, it is used commercially as a sweetener in baking and products that need to have a long shelf life. It is also used in combination with other sweeteners in carbonated drinks and is typically used as the main sweetening agent in pharmaceutical products.

Chemical Identifiers

CAS 55589-62-3
Molecular Formula C4H4KNO4S
Molecular Weight (g/mol) 201.24
InChI Key JLEKLYQXZHJOTQ-UHFFFAOYSA-M
IUPAC Name potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
SMILES [K+].CC1=CC(=O)N=S([O-])(=O)O1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 99%
Grade Reagent