missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame K 98.0+%, TCI America™

Catalog No. A149025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A149025G 25 g
A1490100G 100 g
A1490500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A149025G Supplier TCI America Supplier No. A149025G

Chemical Identifiers

CAS 55589-62-3
Molecular Formula C4H4KNO4S
Molecular Weight (g/mol) 201.237
MDL Number MFCD00043833
InChI Key WBZFUFAFFUEMEI-UHFFFAOYSA-M
Synonym 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-Dioxide Potassium Salt, Potassium 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-Dioxide
PubChem CID 11074431
IUPAC Name potassium;6-methyl-2,2-dioxo-1-oxa-2$l^{6}-thia-3-azanidacyclohex-5-en-4-one
SMILES CC1=CC(=O)[N-]S(=O)(=O)O1.[K+]

Specifications

Color White
Quantity 25 g
Formula Weight 201.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acesulfame K