missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame K, For Food Analysis, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014601
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014601 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014601 Supplier Merck Emd Millipore Supplier No. 04054100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55589-62-3
Molecular Formula C4H4KNO4S
Molecular Weight (g/mol) 201.24
MDL Number MFCD00043833
InChI Key JLEKLYQXZHJOTQ-UHFFFAOYSA-M
Synonym 6-Methyl-1,2,3-oxathiazin-4(3 H)-one 2,2-dioxide potassium salt
IUPAC Name potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
SMILES [K+].CC1=CC(=O)N=S([O-])(=O)O1

Specifications

Linear Formula C4H4KNO4S
Quantity 100 g
Formula Weight 201.24
Percent Purity ≥99.0% (HPLC)
Grade For food analysis