missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthenequinone 98.0+%, TCI America™

Catalog No. A000425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A000425G 25 g
A0004100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A000425G Supplier TCI America Supplier No. A000425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82-86-0
Molecular Formula C12H6O2
Molecular Weight (g/mol) 182.178
MDL Number MFCD00003805
InChI Key AFPRJLBZLPBTPZ-UHFFFAOYSA-N
Synonym acenaphthenequinone, acenaphthoquinone, 1,2-acenaphthylenedione, acenaphthenedione, 1,2-acenaphthenequinone, 1,2-acenaphthenedione, acenaphthaquinone, acenaphthylene-1,2-quinone, acenaphthylenequinone, acenaphthodione
PubChem CID 6724
ChEBI CHEBI:15342
IUPAC Name acenaphthylene-1,2-dione
SMILES C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2

Specifications

Melting Point 255°C
Quantity 25 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acenaphthenequinone
RTECSNumber AB1024500
TSCA Yes