missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthenequinone, 95%

Catalog No. AC102191000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102191000 100 g Glass bottle
AC102190250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102191000 Supplier Thermo Scientific Chemicals Supplier No. 102191000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82-86-0 | C12H6O2 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82-86-0
Molecular Formula C12H6O2
Molecular Weight (g/mol) 182.18
MDL Number MFCD00003805
InChI Key AFPRJLBZLPBTPZ-UHFFFAOYSA-N
Synonym acenaphthenequinone, acenaphthoquinone, 1,2-acenaphthylenedione, acenaphthenedione, 1,2-acenaphthenequinone, 1,2-acenaphthenedione, acenaphthaquinone, acenaphthylene-1,2-quinone, acenaphthylenequinone, acenaphthodione
PubChem CID 6724
ChEBI CHEBI:15342
IUPAC Name acenaphthylene-1,2-dione
SMILES C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 257.0°C to 263.0°C
Color Ochre
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 07, 744
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,benzene,soluble in ligroin
Formula Weight 182.18
Percent Purity 95%
Physical Form Powder or Granules
Chemical Name or Material Acenaphthenequinone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-441-3

RUO – Research Use Only