Learn More
Acenaphthenequinone, 95%
Description
Acenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.
Solubility
Soluble in chloroform and alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 253°C to 262°C |
| Quantity | 5 g |
| Beilstein | 879172 |
| Solubility Information | Soluble in chloroform and alcohol. Insoluble in water. |
| Formula Weight | 182.18 |
| Percent Purity | 95% |
| Chemical Name or Material | Acenaphthenequinone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 201-441-3 |
| RTECSNumber | AB1024500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only