missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthene 99.0+%, TCI America™

Catalog No. p-7116650
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A000325G 25 g
A0003100G 100 g
A0003500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A000325G Supplier TCI America Supplier No. A000325G
Only null left

Chemical Identifiers

CAS 83-32-9
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00003807
InChI Key CWRYPZZKDGJXCA-UHFFFAOYSA-N
Synonym acenaphthene, 1,8-ethylenenaphthalene, naphthyleneethylene, peri-ethylenenaphthalene, acenaphthylene, 1,2-dihydro, ethylenenaphthalene, 1,8-dihydroacenaphthalene, unii-v8ut1gac5y, ccris 5951, v8ut1gac5y
PubChem CID 6734
ChEBI CHEBI:22154
IUPAC Name 1,2-dihydroacenaphthylene
SMILES C1CC2=C3C1=CC=CC3=CC=C2

Specifications

Melting Point 94°C
Color White
Boiling Point 279°C
Quantity 25 g
UN Number 3077
Formula Weight 154.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acenaphthene
EINECSNumber (4)-0645
RTECSNumber AB1000000
TSCA Yes