missing translation for 'onlineSavingsMsg'
Learn More

Acebutolol hydrochloride

Catalog No. AAJ6379009
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6379009 10 g
AAJ6379003 1 g
AAJ6379006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6379009 Supplier Thermo Scientific Chemicals Supplier No. J6379009

Cardioselective beta-adrenergic blocker | CAS: 34381-68-5 | C18H29ClN2O4 | 372.89 g/mol

Cardioselective beta-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34381-68-5
Molecular Formula C18H29ClN2O4
Molecular Weight (g/mol) 372.89
MDL Number MFCD00078860
InChI Key KTUFKADDDORSSI-UHFFFAOYNA-N
Synonym acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn
PubChem CID 441307
ChEBI CHEBI:2380
IUPAC Name hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride
SMILES [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O

Specifications

Melting Point 141°C to 143°C
Color White
Quantity 10 g
Merck Index 14,19
Formula Weight 372.89
Physical Form Powder
Chemical Name or Material Acebutolol hydrochloride
EINECSNumber 251-980-3
RTECSNumber ES5235000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only