missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AccuSEQ™ Real-Time PCR Detection Software v3.1

Catalog No. 361055936
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361055936 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361055936 Supplier Applied Biosystems™ Supplier No. A48509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Content And Storage Product letter and License registration code kit
For Use With (Equipment) QuantStudio™ 5, 0.1 mL Block Instrument
Product Line AccuSEQ
Software Type Real-Time PCR Software
Product Type Real Time PCR Detection Software
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Room Temperature
Quantity 1 Kit
When using the AccuSEQ Real-Time PCR Detection Software v3.1 (Cat. No. A48509), for the double signature function, do the users who will sign have to have different roles or could they also hold the same role?

With the AccuSEQ Real-Time PCR Detection Software v3.1 (Cat. No. A48509), the administrator can define whether 1 or 2 signatures are required to sign an experiment. However, it is not possible to specify that the signatures have to come from different roles. The administrator can only define how many signatures (up to 10) are required to sign an experiment.

For Research Use Only. Not for use in diagnostic procedures.