missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Gel-Filled Paper Pulp Double-Junction pH Combination Electrodes: Mercury-Free

Catalog No. 13620299A
Change view
Click to view available options
Connector Types:
BNC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13620299A BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13620299A Supplier Fisherbrand™ Supplier No. 13 620 299A
Only null left

Dual junctions provide fast response and long life in paper pulp testing

  • Perfect for paper pulp testing, mine assay labs, smelters—anywhere junction clogging may be a problem
  • Inner chamber houses reference element with gelled KCl and AgCl; the outer chamber contains KCl only
  • Eliminates heavy metal ions that react with sulfide ions in samples such as black and green liquors
  • No insoluble sulfide precipitates to block electrode junction and cause premature failure
  • Polypropylene body
  • Fisher Scientific universal glass membrane for rapid, reproducible response
  • Glass tip is recessed in end
Fast and accurate
  • Accuracy, ±0.05pH at 25°C (77°F) Efficiency, >95% Sodium Ion error, <±0.05pH at 25°C (77°F) Response time, 30 seconds Zero potential, 7.0 ±0.3pH
  • Membrane resistance, <100megohms at 25°C (77°F)

  • Ideal for routine process monitoring in paper pulp plants, mine assay labs, smelters
  • Fits any application where the presence of S= in samples causes problems of slow response and junction failure

Specifications

Connector Types BNC
Junction Dual
Material Gel-filled Polymer/Polypropylene
pH Range 0 to 14
Type Combination pH Electrode
Filling Solution Type Gel
For Use With (Application) Ideal for routine process monitoring in paper pulp plants, mine assay labs, smelters
For Use With (Equipment) pH meter
pH Relative Accuracy ±0.05
Response Time 30 sec.