Learn More
Fisherbrand™ accumet™ Four Cell Conductivity/ATC Probes, Four-Cell
For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters
Supplier:  Fisherbrand™ 13 620 163
 View more versions of this product
Catalog No. 13620163
Only null left
Description
Description
For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters
Immersion-type cell's unique compact size is the same as a typical pH electrode—allowing use in standard electrode holder.
  • Four-cell type with glass or epoxy bodies
  • Built-in temperature sensors and 8-pin connectors
  • Cell constants of 0.1, 1.0, or 10.0cm-1
Specifications
Chemical Identifiers
Conductivity/ATC Probe
8-pin
Immersed in deionized water
4 Cell Conductivity
Specifications
1 cm-1
Fisher Scientific accumet benchtop models XL600, XL500, XL200, & AB200 and legacy models XL60, XL50, XL30, XL20, AR60, AR50, AR30, AR20, & AB30
Glass
Immersed in deionized water
Conductivity/ATC Probe
8-pin
10 μS to 100 mS/cm
4 Cell Conductivity
Product Suggestions
Product Suggestions