missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AR415 pH Bench Meter

Catalog No. 13636AR415
Change view
Click to view available options
Model:
AR415
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
13636AR415 AR415
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13636AR415 Supplier Fisherbrand™ Supplier No. 13636AR415
Get a pH Buffer Kit with pH/mV Meter Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to deliver precise and reliable electrochemistry testing, the accumet AR415 offers enhanced performance in a streamlined design. Ideal for laboratories requiring dependable measurement capabilities.

  • Large 7-in. high-resolution color display with capacitive touchscreen operable with gloves.
  • One to five point pH calibration with automatic buffer recognition.
  • Streamlined, easy-to-operate user interface with step-by-step calibration guidance.
  • Stores 10 calibration logs per parameter with date and time.
  • Stores 5000 data log sets with date and time.
  • Outputs to a printer, computer, or USB flash drive for long-term data retention.
  • Inputs sample ID with optional USB barcode scanner, USB keyboard, or USB mouse.
  • Stirs samples with meter-controlled and meter-powered stirrer probe.

Specifications

Accuracy (Temperature) ±0.3°C
Accuracy (pH) ±0.002 pH
Includes pH Meter
Accuracy (mV) -2000.0 to 2000.0 mV
Measurement Modes pH, ORP, mV, Temperature
Calibration pH, Relative mV, Temperature
Range (pH) -2 to 20
Resolution (pH) 0.1, 0.01, or 0.001 pH
Range (mV) -2000.0 to 2000.0 mV
Calibration Points 1
Temperature Range (Metric) -30.0 to +130.0°C
Display Type 7-in. High-resolution Color Graphic LCD
Backlight Option Yes
Data Points 5000 with Date and Time Stamp
Log Function Type Automatic Data Logging
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP54
Model AR415
Channels 1
Type pH Meter
Voltage 100/240 V
Show More Show Less