Learn More
Ammonia pH/ISA, 10 M Sodium Hydroxide, Fisherbrand
Ammonia pH/Ionic Strength Adjuster
Supplier: Ricca Chemical Company ACCU0802500A
Description
Ammonia pH/Ionic Strength Adjuster to eliminate interference from other ions
- pH/Ionic strength adjuster
Specifications
| Ammonia pH/Ionic Strength Adjuster | |
| pH/Ionic Strength Adjuster | |
| 500 mL |
| 10 M NaOH | |
| Natural Poly Bottle |