Learn More
Accuform Signs Self-Adhesive Individual Numerals
Use numbers and letters to properly mark and identify objects and areas.
Supplier:  Accuform Signs NAK1012
 View more versions of this product
Catalog No. 19805030
Only null left
Description
Description
Specifications
Chemical Identifiers
Vinyl
Multitude of identification needs in various environments
176°F
-20°C to 80°C
Individual Number
50/Pk.
3.5 in.
-20°F
-20°F to 176°F
Black on White
Specifications
Vinyl
Multitude of identification needs in various environments
176°F
-20°C to 80°C
2
Black on White
50/Pk.
3.5 in.
-20°F
-20°F to 176°F
Individual Number
Product Suggestions
Product Suggestions