missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Safety Signs
Description
Keep employees safe from harm by effectively communicating hazardous situations to others
- Warn employees not to touch the surface of equipment and reduce workplace injuries.
- Caution others so they're aware of hot surfaces in the work area.
Specifications
Specifications
| Material | Vinyl |
| Description | Dura Vinyl |
| Height (English) | 3.5 in. |
| Max. Temperature (English) | 176°F |
| Min. Temperature (English) | -20°F |
| Width (English) | 5 in. |
| Temperature Range (Metric) | -29°C to +80°C |
| Temperature Range (English) | -20°F to +176°F |
| Dimensions (L x W) | 3.5 x 5 in. |
| Displayed Text | CAUTION Hot Surface |