missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Safety Signs

Catalog No. 19805301
Change view
Click to view available options
Material:
Aluminum
Polyethylene
Vinyl
Displayed Text:
CAUTION
CAUTION Hot Surface
DANGER Hot Do Not Touch
Product Type:
Safety Sign
Safety Tape
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Displayed Text Product Type
19805301 Aluminum DANGER Hot Do Not Touch Safety Sign
19805302 Vinyl CAUTION Hot Surface -
19805303 Polyethylene CAUTION Safety Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19805301 Supplier Accuform Signs Supplier No. PAT725
Only null left

Keep employees safe from harm by effectively communicating hazardous situations to others

Specifications

Material Aluminum
Height (English) 3.5 in.
Max. Temperature (English) 140°F
Min. Temperature (English) -20°F
Width (English) 5 in.
Temperature Range (Metric) -28°C to +60°C
Temperature Range (English) -20°F to +140°F
Displayed Text DANGER Hot Do Not Touch
Product Type Safety Sign