Learn More
Accuform Signs Retro-Reflective Helmet Stickers
Catalog No. 19804586
Change view
Click to view available options
Color:
Fluorescent Lime Green, Yellow
Fluorescent Red, Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19804586 Fluorescent Lime Green, Yellow
19804587 Fluorescent Red, Orange
19809843 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19804586 Supplier Accuform Signs Supplier No. LHR104GNYL
Easily identify workers in dark conditions with these highly reflective helmet stickers.
  • Helmet stickers are reflective, so workers can be seen in a wide range of light conditions
  • Stickers have a self-adhesive back with a peel-off liner for easy application
  • 14-mil vinyl contains microprism pattern
Specifications
Color Fluorescent Lime Green, Yellow