missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Non-Hazardous-Waste Labels

Catalog No. 19804831
Change view
Click to view available options
Color:
White, Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19804831 White, Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19804831 Supplier Accuform Signs Supplier No. MHZW11PSL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use these labels when identifying waste that is non-hazardous and does not pose a threat.

Specifications

Certifications/Compliance DOT (49 CFR Part 172) and EPA (40 CFR Part 262.31 - 32)
Adhesive Type Self-adhesive
Color White, Green
Material Paper
Product Type Non-Hazardous Waste Label
Display Text Non Hazardous Waste Optional Information Shipper Address City, State, Zip Contents
For Use With (Application) For labeling substance containers for hazardous waste
Quantity 250/Pk.
Width (English) 6 in.
Height (English) 6 in.
Height (Metric) 15.24 cm
Show More Show Less