missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs NFPA Placards

Catalog No. 19121451
Change view
Click to view available options
Material:
Aluminum
Vinyl
Height (Metric):
10.2 cm
15.2 cm
25.4 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (Metric)
19121451 Vinyl 10.2 cm
19121453 Vinyl 15.2 cm
19121452 Aluminum 25.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19121451 Supplier Accuform Signs Supplier No. ZFD404EV

Designed for on-the-spot classification of potentially hazardous chemicals

  • Printed with proper color coding
  • UV stable inks
  • Indiviudal, pre-printed hazard panels are available separately
  • Available as blank placards or as part of a kit

Placard Kit: (Nos 19-121-451 and 19-121-453)

  • 15 numbers (3 sets of 0-4)
  • 7 specific-hazard symbols (Use No Water, Oxidizer, Simple Asphyxiant, Radiation Hazard, Corrosive, Acid and Alkali)
  • NFPA Wallet Guide
  • Blank Placard

Specifications

Includes 15 Numbers (3 sets of 0 to 4), 7 specific hazard symbols (Use No Water, Oxidizer, Simple Asphyxiant, Radiation Hazard, Corrosive, Acid and Alkali), NFPA wallet guide, blank placard
Material Vinyl
Quantity 1 Ea.
Mounting Type Surface Mount
Height (English) 4 in.
Height (Metric) 10.2 cm
Thickness 2 mil
Width (English) 4 in.
Width (Metric) 10.2 cm
Shape Diamond
Product Type NHPA Placard Kit
Color Blue
Show More Show Less