missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs NFPA Diamond Roll Labels
Quickly identify any serious hazards present associated with the chemicals.
Supplier: Accuform Signs LZN635EV5
Chemical Identifiers
| Self-adhesive | |
| Hand-written hazard codes and symbols | |
| NFPA Label | |
| White |
Specifications
| Polyester | |
| 6 x 6 in. | |
| Self-adhesive | |
| -40°C to +121°C | |
| NFPA Label |
| 500/Pk. | |
| 15.2 x 15.2 cm | |
| Hand-written hazard codes and symbols | |
| -40°F to +250°F | |
| White |