missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs NFPA Diamond Roll Labels
Specifications
Specifications
| Material | Paper |
| Quantity | 500/Pk. |
| Size (English) | 4 x 4 in. |
| Size (Metric) | 10.1 x 10.1 cm |
| Mounting Type | Self-adhesive |
| For Use With (Application) | Hand-written hazard codes and symbols |
| Product Type | NFPA Label |
| Color | White |