missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs NFPA Diamond Roll Labels
Quickly identify any serious hazards present associated with the chemicals.
Supplier: Accuform Signs LZN631PS5
Chemical Identifiers
| Self-adhesive | |
| Hand-written hazard codes and symbols | |
| NFPA Label | |
| White |
Specifications
| Paper | |
| 2 x 2 in. | |
| Self-adhesive | |
| NFPA Label |
| 500/Pk. | |
| 5.08 x 5.08 cm | |
| Hand-written hazard codes and symbols | |
| White |