missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs NFPA Classification Roll Labels
Description
To ensure the quick identification of fire hazards caused by chemicals
- Classifies what code each number represented corresponds to, to indicate the danger level.
- Identifier for chemicals commonly used or encountered in facilities or factories.
- Sticker features NFPA diamond identifier to label potentially hazardous chemicals.
- Inform readers of the details of a hazardous chemical with the appropriate information.
Specifications
Specifications
| Certifications/Compliance | National and global agencies |
| Material | Paper |
| Quantity | 500/Pk. |
| Size (English) | 1.5 x 3.875 in. |
| Size (Metric) | 3.81 x 9.84 cm |
| Mounting Type | Self-adhesive |
| For Use With (Application) | Chemical identification |
| Dimensions (L x W) | 1.5 x 3.875 in. |
| Product Type | NFPA Label |