missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs NFPA Classification Roll Labels

to ensure the quick identification of fire hazards caused by chemicals

Supplier:  Accuform Signs LZN601EV

 View more versions of this product

Catalog No. 19804607

Only null left

Description

Description

Specifications

Chemical Identifiers

500/Pk.
Self-adhesive

Specifications

Polyester
1.5 x 3.875 in.
Self-adhesive
2.6 mil
-40°F to +250°F
NFPA Label
500/Pk.
3.81 x 9.84 cm
Chemical identification
-40°C to +121°C
1.5 x 3.875 in.
White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions