missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Machinery and Operations Labels
Description
Place Automatic Equipment Labels throughout your facility to effectively communicate information that keeps workers safe while working with and around specific machinery.
- Work Above and Below signs warn people about a variety of equipment hazards either above their head, or below their feet.
- Prevent injuries by mandating that safety guards are used when operating machinery.
- Remind workers to keep hands away from hazards with a well-placed safety label.
- Ensure safety protocols are exercised by mandating the use of machine guards.
Specifications
Specifications
| Material | Vinyl |
| Quantity | 1 Ea. |
| Size (English) | 3.5 x 5 in. |
| Size (Metric) | 8.89 x 12.7 cm |
| Mounting Type | Self-adhesive |
| Temperature Range (Metric) | -20°C to +80°C |
| Temperature Range (English) | -20°F to +176°F |
| Displayed Text | Caution Keep Machine Guards In Place |
| Product Type | Equipment Safety Label |
| Life Span | 5 Years |