Learn More
Accuform Signs Machinery and Operations Labels
Place Automatic Equipment Labels throughout your facility to effectively communicate information that keeps workers safe while working with and around specific machinery.
Supplier:  Accuform Signs LEQM615XVE
 View more versions of this product
Catalog No. 19-804-578
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Vinyl
3.5 x 5 in.
Self-adhesive
-20°F to +176°F
Equipment Safety Label
1 Ea.
8.89 x 12.7 cm
-20°C to +80°C
Caution Do Not Operate This Machine Without Guard In Place
5 Years
Product Suggestions
Product Suggestions