missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Hazardous Materials Identification Index Poster

Catalog No. 19804523
Change view
Click to view available options
Dimensions (L x W):
18 x 24 in. (45.7 x 61 cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19804523 18 x 24 in. (45.7 x 61 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19804523 Supplier Accuform Signs Supplier No. SP125164L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Poster for quick look-up of the hazard index and required personal protection

  • Easy to read legend utilizes pictorials to indicate which personal protective equipment is necessary for a specific code
  • Size: 45.7 × 61cm (18 × 24 in.)
  • Colorful and highly visible
  • Choose between English and Spanish languages
  • Available in an encapsulated laminate bond

Specifications

Material Laminated Plastic
For Use With (Application) Quick look up of the hazard index and personal protection
Width (English) 24 in.
Width (Metric) 61 cm
Length (English) 18 in.
Length (Metric) 45.7 cm
Dimensions (L x W) 18 x 24 in. (45.7 x 61 cm)
Product Type Hazardous Materials Identification Index Poster
Color Multi-colored