Learn More
Accuform Signs Glow-in-the-Dark Marking Tapes
Catalog No. 19805184
Change view
Click to view available options
Color:
Green, Yellow
Catalog No. Color
19805184 Green, Yellow
1 options
Catalog No. 19805184 Supplier Accuform Signs Supplier No. PTM912
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mark for visibility and safety in low light and emergency blackout situations
  • Size: 0.05cm (2 in.) x 9.14m (30 ft.)
  • Photo-luminescent marking tapes glow in the dark in a green-yellow color
  • Marking tape is energized by any light source
  • Longer exposure to increases glow time
  • Good marking visibility in regular light shows white with slight green appearance
  • Made of flexible 10-mil polyester with laminated gloss surface and permanent Acrylic adhesive back with peel-off liner
Specifications
Dimensions (L x W) 30 ft. x 2 in.
Color Green, Yellow