missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Electrical Safety Labels
Description
Provide information about potential hazards to keep employees safe when using electrical equipment.
- Warn employees of the dangers of electric shock from 480 volt breaker boxes.
- Warn of the danger of high voltage areas to prevent electrical injuries.
- Alert employees and guests of potential electrical hazards in the immediate vicinity.
Specifications
Specifications
| Certifications/Compliance | OSHA 29 CFR 1910.145 |
| Finish | Non-glare |
| Material | Vinyl |
| Quantity | 5/Pk. |
| For Use With (Application) | Provide proper protection in certain areas in order to stay safe on the job or in the for short term outdoor and self sticking applications |
| Height (English) | 3.5 in. |
| Max. Temperature (English) | 176°F |
| Min. Temperature (English) | -20°F |
| Thickness | 4 mil |
| Width (English) | 5 in. |
| Show More |