missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Danger: Compressed Air Sign
Description
Alert others to the presence of compressed air in the work environment.
- Inform all personnel of the presence of compressed air and its corresponding hazards.
- Alert others to the presence of compressed air in the work environment.
Specifications
Specifications
| Certifications/Compliance | ANSI, OSHA |
| Material | Plastic |
| For Use With (Application) | Marking content of a cylinder, identifying cylinder group, use status and safety message |
| Height (English) | 10 in. |
| Min. Temperature (English) | -90°F |
| Thickness | 0.06 in. |
| Width (English) | 14 in. |
| Temperature Range (Metric) | -68°C to +76°C |
| Temperature Range (English) | -90°F to +168°F |
| Displayed Text | DANGER Compressed Air |
| Show More |