missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Confined Space Signs

Catalog No. 19804693
Change view
Click to view available options
Dimensions (L x W):
25.4 x 35.6cm (10 x 14 in.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19804693 25.4 x 35.6cm (10 x 14 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19804693 Supplier Accuform Signs Supplier No. MCSP002VS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Colorful, compact safety signs

  • Material: Adhesive vinyl
  • Color: White, Black, Red
  • Size: 25.4 × 35.6cm (10 × 14 in.)
  • Language: English

Specifications

Certifications/Compliance 29 CFR 191.146
Material Vinyl
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage
Height (English) 10 in.
Height (Metric) 25.4 cm
Max. Temperature (English) 146°F
Min. Temperature (English) -20°F
Thickness 4 mil
Width (English) 14 in.
Width (Metric) 35.6 cm
Temperature Range (Metric) -29°C to 63°C
Temperature Range (English) -20°F to 146°F
Dimensions (L x W) 25.4 x 35.6cm (10 x 14 in.)
Displayed Text DANGER Confined Space
Product Type Confined Space Sign
Color Red on White
Show More Show Less