Learn More
Accuform Signs Chemical Labels
for effectively communicating hazardous contents by placing proper labeling and Chemical Identification in and around your facility.
Supplier:  Accuform Signs LCHL532
 View more versions of this product
Catalog No. 19804566
Only null left
Description
Description
Specifications
Chemical Identifiers
OSHA 29 CFR 1910.145
1 Ea.
Communicating the potential dangers posed by chemicals either in storage or in use
-20°C to +80°C
Chemical and Hazardous Safety Label
Vinyl
Self-adhesive
6 mil
-20°F to +176°F
5 to 7 Years
Specifications
OSHA 29 CFR 1910.145
1 Ea.
Communicating the potential dangers posed by chemicals either in storage or in use
-20°C to +80°C
Diesel Fuel
Black, White
Vinyl
Self-adhesive
6 mil
-20°F to +176°F
Chemical and Hazardous Safety Label
5 to 7 Years
Product Suggestions
Product Suggestions