missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Caution Watch Your Step Foldup Sign

Catalog No. 19804323
Change view
Click to view available options
Dimensions (L x W):
20 x 12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19804323 20 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19804323 Supplier Accuform Signs Supplier No. MF110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Free-standing sign for hazardous footing

  • Legend: CAUTION. WATCH YOUR STEP (w/ graphic)
  • Material: .125 in. high-impact plastic
  • Size (H × W): 50.8 × 30.5cm (20 × 12 in.)
  • Handle cut-out for ease of carrying
  • Folds flat for transport and storage
  • Both sides have same message
  • Color: Yellow, Black
  • Language: English

Specifications

Material Plastic
Quantity 1 Ea.
For Use With (Application) Identifying temporary hazardous conditions or displaying important messages, quickly and easily
Height (English) 20 in.
Height (Metric) 50.8 cm
Max. Temperature (English) 168°F
Min. Temperature (English) -90°F
Thickness 0.06 in.
Width (English) 12 in.
Width (Metric) 30.5 cm
Temperature Range (Metric) -68°C to +76°C
Temperature Range (English) -90°F to +168°F
Languages English
Dimensions (L x W) 20 x 12 in.
Displayed Text CAUTION Watch Your Step
Product Type Safety Sign
Color Red on Yellow
Show More Show Less