missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Biohazard Signs
Description
- Legend: CAUTION BIOHAZARD (w/ graphic)
- Size: 35.6 × 25.4cm (14 × 10 in.)
- Material: Plastic
- Color: Orange, Black
- Language: English
Specifications
Specifications
| Certifications/Compliance | ASIN B000RMJ3WW |
| Material | Plastic |
| Quantity | 1 Ea. |
| Height (English) | 10 in. |
| Height (Metric) | 35.6 cm |
| Width (English) | 14 in. |
| Width (Metric) | 25.4 cm |
| Dimensions (L x W) | 14 x 10 in. |
| Displayed Text | CAUTION Biohazard |
| Product Type | Biohazard Sign |
| Show More |