missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bioscience accu-Test™ Vials for COD Testing
For low-, standard- and high-range concentrations
Supplier: Bioscience 174318
Includes: 16mm vial with 1.5mL digestion solution, 3.5mL catalyst solution, and screw cap
Description
- Convenient to use
- Give excellent analytical results
- Compatible with all standard COD reactors and spectrophotometers
- For samples with low chloride and ammonia levels, mercury-free reagents are available for nonreporting purposes
Order Info
Supplied in modular packs that are easy to stack
Certifications
U.S. EPA approved (for standard and low ranges) micro Chemical Oxygen Demand test
Recommended Storage
Shelf Life: Minimum of one year from date of manufacture
Specifications
| EPA, OSHA | |
| COD testing for 20 to 900mg/L standard range concentration | |
| Mercuric Sulfate |
| 75% Sulfuric Acid, 0.20% Potassium Dichromate, 0.40% Silver Sulfate, 0.002%Sulfamic Acid, 0.60% Mercuric Sulfate | |
| Standard COD reactor and spectrophotometer | |
| 20 to 900 mg/L |