missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Accessory for Titrator: Injection Needles

Catalog No. 01911235
Change view
Click to view available options
Type:
80 x 0.8 mm Injection Needles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01911235 80 x 0.8 mm Injection Needles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01911235 Supplier Mettler Toledo™ Supplier No. 71484

For use with D32 Karl Fischer titrator

Injection needle: 80 x 0.8mm

Specifications

Type 80 x 0.8 mm Injection Needles
For Use With (Application) Titration
Quantity 100 Pk.
Size 80 x 0.8mm
For Use With (Equipment) D32 Karl Fischer titrator