Learn More
BD Accessories for Veritor™ Plus Analyzer
Catalog No. b443907
Change view
Click to view available options
Type:
InfoScan Module
Printer Cable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
B443907 Printer Cable
B256068 InfoScan Module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B443907 Supplier BD Supplier No. 443907
Accessories for use with Veritor Plus Analyzer.
InfoScan Module
  • Point of Care scanning of three different identifiers - Operator ID, Specimen ID, and Test Kit/Lot information
  • Identifiers can be locally recorded, printed or downloaded and are automatically documented
  • Barcode scanner
  • Facilitate traceability and help reduce manual transcription
  • Data access in a CSV file downloaded via USB to a local computer or print-enabled
Printer Cable
  • Custom printer cable to connect to Epson TM-M30, TM-L90 or TM-M20II
Specifications
Type Printer Cable
Description For use with Veritor Plus Analyzer